MAN372903986 Manchester Organics

1,4-Dimethoxy-2-fluorobenzene

  • MDL number MFCD00042487
  • Cas Number : 82830-49-7
  • Molecular Weight : 156.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/K23830
Availability details for
Quantity
  • Availability
  • Add to Cart