MAN372904183 Manchester Organics

(R)-Piperidin-3-ylmethyl-carbamic acid tert-butyl ester

  • MDL number MFCD03093384
  • Cas Number : 879275-33-9
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/K25392
Availability details for
Quantity
  • Availability
  • Add to Cart