MAN372904184 Manchester Organics

S-Piperidin-3-ylmethyl-carbamic acid tert-butyl ester

  • MDL number MFCD03093385
  • Molecular Weight : 214.305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L25393
Availability details for
Quantity
  • Availability
  • Add to Cart