MAN372904222 Manchester Organics

N-Boc-3-Methylpiperidin-4-one

  • MDL number MFCD08460962
  • Molecular Weight : 213.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E25412
Availability details for
Quantity
  • Availability
  • Add to Cart