MAN372904235 Manchester Organics

2,2,3,4,4,4-Hexafluorobutanoic acid

  • MDL number MFCD16621311
  • Cas Number : 379-90-8
  • Molecular Weight : 196.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G25224
Availability details for
Quantity
  • Availability
  • Add to Cart