MAN372904301 Manchester Organics

Trimethylpyruvic acid

  • MDL number MFCD00154352
  • Cas Number : 815-17-8
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/A25454
Availability details for
Quantity
  • Availability
  • Add to Cart