MAN372904378 Manchester Organics

Diethyl 2,6-dibromoheptanedioate

  • MDL number MFCD00087327
  • Cas Number : 868-68-8
  • Molecular Weight : 374.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl 2,6-dibromoheptanedioate
Inchi Code 1S/C11H18Br2O4/c1-3-16-10(14)8(12)6-5-7-9(13)11(15)17-4-2/h8-9H,3-7H2,1-2H3
Inchi Key SPJSPERKKWTTBF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/L12164
Availability details for
Quantity
  • Availability
  • Add to Cart