MAN372904525 Manchester Organics

2,3-Dihydro-1,4-benzodioxin-6yl-methylamine

  • MDL number MFCD02677684
  • Cas Number : 17413-10-4
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/F25743
Availability details for
Quantity
  • Availability
  • Add to Cart