MAN372904545 Manchester Organics

2,4-Dichloro-6-methylpyridine

  • MDL number MFCD08669834
  • Cas Number : 42779-56-6
  • Molecular Weight : 162.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/D24014
Availability details for
Quantity
  • Availability
  • Add to Cart