MAN372904555 Manchester Organics

3-Amino-6-bromopyridazine

  • MDL number MFCD01646092
  • Molecular Weight : 174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A25761
Availability details for
Quantity
  • Availability
  • Add to Cart