MAN372904595 Manchester Organics

(Z)-Benzyl(1H-pyrazol-1-yl)methylenedicarbamate

  • MDL number MFCD02092983
  • Cas Number : 152120-55-3
  • Molecular Weight : 378.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G25485
Availability details for
Quantity
  • Availability
  • Add to Cart