MAN372904604 Manchester Organics

6-Nitrooxindole

  • MDL number MFCD09835636
  • Cas Number : 474799-41-2
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D24037
Availability details for
Quantity
  • Availability
  • Add to Cart