MAN372904608 Manchester Organics

Oxetane-2-carboxylic acid

  • MDL number MFCD08544406
  • Cas Number : 864373-47-7
  • Molecular Weight : 102.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/E24038
Availability details for
Quantity
  • Availability
  • Add to Cart