MAN372904632 Manchester Organics

2-Amino-6-chloro-3-pyridinecarboxamide

  • MDL number MFCD12024435
  • Cas Number : 64321-24-0
  • Molecular Weight : 171.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/V12172
Availability details for
Quantity
  • Availability
  • Add to Cart