MAN372904641 Manchester Organics

2,6-Dibromopyrazine

  • MDL number MFCD09834804
  • Cas Number : 23229-25-6
  • Molecular Weight : 237.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/S24049
Availability details for
Quantity
  • Availability
  • Add to Cart