MAN372904695 Manchester Organics

4-Chloro-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD04972771
  • Cas Number : 37552-81-1
  • Molecular Weight : 182.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-6-(trifluoromethyl)pyrimidine
Inchi Code 1S/C5H2ClF3N2/c6-4-1-3(5(7,8)9)10-2-11-4/h1-2H
Inchi Key TYSPDLZOMUDHQZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/F24087
Availability details for
Quantity
  • Availability
  • Add to Cart