MAN372904712 Manchester Organics

4-Bromo-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD10574962
  • Cas Number : 785777-89-1
  • Molecular Weight : 226.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/F23542
Availability details for
Quantity
  • Availability
  • Add to Cart