MAN372904736 Manchester Organics

2-Chloropyrimidine-5-carbaldehyde

  • MDL number MFCD10696891
  • Cas Number : 933702-55-7
  • Molecular Weight : 142.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/V25520
Availability details for
Quantity
  • Availability
  • Add to Cart