MAN372904756 Manchester Organics

2,6-Dimethyl-3,5-dibromopyridine

  • MDL number MFCD00234282
  • Cas Number : 3430-34-0
  • Molecular Weight : 264.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/G24111
Availability details for
Quantity
  • Availability
  • Add to Cart