MAN372904778 Manchester Organics

5-Chloro-2-hydroxypyrazine

  • MDL number MFCD11109884
  • Cas Number : 89180-45-0
  • Molecular Weight : 130.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/U24122
Availability details for
Quantity
  • Availability
  • Add to Cart