MAN372904794 Manchester Organics

(3S)-3-(Trifluoromethyl)piperidine

  • MDL number MFCD11975307
  • Cas Number : 749208-62-6
  • Molecular Weight : 153.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M15454
Availability details for
Quantity
  • Availability
  • Add to Cart