MAN372904813 Manchester Organics

2-(Piperidin-4-yloxy)pyrimidine dihydrochloride

  • MDL number MFCD03840123
  • Cas Number : 950649-19-1
  • Molecular Weight : 252.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/S25824
Availability details for
Quantity
  • Availability
  • Add to Cart