MAN372904817 Manchester Organics

(3-Trifluoromethylphenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD11040633
  • Cas Number : 109134-07-8
  • Molecular Weight : 261.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(trifluoromethyl)phenylcarbamate
Inchi Code 1S/C12H14F3NO2/c1-11(2,3)18-10(17)16-9-6-4-5-8(7-9)12(13,14)15/h4-7H,1-3H3,(H,16,17)
Inchi Key PLLZDOLFXQKWQG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/W25828
Availability details for
Quantity
  • Availability
  • Add to Cart