MAN372904837 Manchester Organics

2-(Trifluoromethyl)acrylonitrile

  • MDL number MFCD01673877
  • Molecular Weight : 121.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B24131
Availability details for
Quantity
  • Availability
  • Add to Cart