MAN372904854 Manchester Organics

1-(5-Chloro-3-methylbenzothiophen-2-yl)ethanone

  • MDL number MFCD00052500
  • Molecular Weight : 224.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/G25863
Availability details for
Quantity
  • Availability
  • Add to Cart