MAN372904857 Manchester Organics

3-Amino-2-cyanopyridine

  • MDL number MFCD02181031
  • Cas Number : 42242-11-5
  • Molecular Weight : 119.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M25536
Availability details for
Quantity
  • Availability
  • Add to Cart