MAN372904859 Manchester Organics

2,6-Dichloro-4-(difluoromethyl)pyridine

  • MDL number MFCD16658727
  • Cas Number : 1201675-01-5
  • Molecular Weight : 198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/N15455
Availability details for
Quantity
  • Availability
  • Add to Cart