MAN372904861 Manchester Organics

3-Chloro-4-(difluoromethyl)pyridine

  • MDL number MFCD22124921
  • Cas Number : 1374659-44-5
  • Molecular Weight : 163.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-4-(difluoromethyl)pyridine
Inchi Code 1S/C6H4ClF2N/c7-5-3-10-2-1-4(5)6(8)9/h1-3,6H
Inchi Key UZRBGKNPGFNLDR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/R15457
Availability details for
Quantity
  • Availability
  • Add to Cart