MAN372904869 Manchester Organics

2,3-Difluoro-4-pyridinecarboxaldehyde

  • MDL number MFCD16610817
  • Molecular Weight : 143.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X15463
Availability details for
Quantity
  • Availability
  • Add to Cart