MAN372904870 Manchester Organics

2,3-Difluoro-4-(difluoromethyl)pyridine

  • MDL number MFCD22199287
  • Cas Number : 1374659-38-7
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(difluoromethyl)-2,3-difluoropyridine
Inchi Code 1S/C6H3F4N/c7-4-3(5(8)9)1-2-11-6(4)10/h1-2,5H
Inchi Key BEKMBPQQOARBLZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y15464
Availability details for
Quantity
  • Availability
  • Add to Cart