MAN372904879 Manchester Organics

5-Methyl-1H-pyrazole

  • MDL number MFCD08685900
  • Molecular Weight : 82.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B25549
Availability details for
Quantity
  • Availability
  • Add to Cart