MAN372904883 Manchester Organics

5-(Difluoromethyl)-2-fluoropyridine

  • MDL number MFCD22199299
  • Molecular Weight : 147.098

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B15467
Availability details for
Quantity
  • Availability
  • Add to Cart