MAN372904887 Manchester Organics

2,3-Dichloro-5-(difluoromethyl)pyridine

  • MDL number MFCD22199295
  • Cas Number : 71690-06-7
  • Molecular Weight : 198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dichloro-5-(difluoromethyl)pyridine
Inchi Code 1S/C6H3Cl2F2N/c7-4-1-3(6(9)10)2-11-5(4)8/h1-2,6H
Inchi Key ODKRLOKESDPEPD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C15470
Availability details for
Quantity
  • Availability
  • Add to Cart