MAN372904892 Manchester Organics

3-(Difluoromethyl)-2-fluoropyridine

  • MDL number MFCD22199298
  • Cas Number : 1374659-42-3
  • Molecular Weight : 147.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/E15472
Availability details for
Quantity
  • Availability
  • Add to Cart