MAN372904895 Manchester Organics

5-Fluoro-3-pyridinecarboxaldehyde

  • MDL number MFCD07778429
  • Cas Number : 39891-04-8
  • Molecular Weight : 125.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/V12195
Availability details for
Quantity
  • Availability
  • Add to Cart