MAN372904900 Manchester Organics

5-Methyl-3-pyridinecarboxaldehyde

  • MDL number MFCD08272078
  • Cas Number : 100910-66-5
  • Molecular Weight : 121.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/X12197
Availability details for
Quantity
  • Availability
  • Add to Cart