MAN372904922 Manchester Organics

2,6-Dichloro-3-(difluoromethyl)-5-fluoropyridine

  • MDL number MFCD22199308
  • Cas Number : 1374659-39-8
  • Molecular Weight : 215.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-dichloro-3-(difluoromethyl)-5-fluoropyridine
Inchi Code 1S/C6H2Cl2F3N/c7-4-2(6(10)11)1-3(9)5(8)12-4/h1,6H
Inchi Key QVEUGAUQNBOLML-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/X15488
Availability details for
Quantity
  • Availability
  • Add to Cart