MAN372904933 Manchester Organics

3-Bromo-2-chloro-5-(difluoromethyl)pyridine

  • MDL number MFCD22199311
  • Cas Number : 71702-02-8
  • Molecular Weight : 242.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/C15493
Availability details for
Quantity
  • Availability
  • Add to Cart