MAN372904946 Manchester Organics

2-(Difluoromethyl)-3,6-difluoropyridine

  • MDL number MFCD22199313
  • Cas Number : 1374659-43-4
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/J15499
Availability details for
Quantity
  • Availability
  • Add to Cart