MAN372904964 Manchester Organics

2-Chloro-3-(difluoromethyl)-6-methylpyridine

  • MDL number MFCD22199315
  • Cas Number : 1374659-36-5
  • Molecular Weight : 177.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N15503
Availability details for
Quantity
  • Availability
  • Add to Cart