MAN372905031 Manchester Organics

4-Bromo-2-fluoro-6-methylaniline

  • MDL number MFCD22683228
  • Cas Number : 429683-46-5
  • Molecular Weight : 204.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W11060
Availability details for
Quantity
  • Availability
  • Add to Cart