MAN372905032 Manchester Organics

5-Bromo-1-fluoro-3-methyl-2-nitrobenzene

  • MDL number MFCD22566379
  • Molecular Weight : 234.023

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X11061
Availability details for
Quantity
  • Availability
  • Add to Cart