MAN372905170 Manchester Organics

1,3-Bis(aminopropyl)tetramethyldisiloxane

  • MDL number MFCD00039786
  • Cas Number : 2469-55-8
  • Molecular Weight : 248.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/G26076
Availability details for
Quantity
  • Availability
  • Add to Cart