MAN372905362 Manchester Organics

3-(Aminosulfonyl)propanoic acid

  • MDL number MFCD09971516
  • Cas Number : 15441-10-8
  • Molecular Weight : 153.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/R26368
Availability details for
Quantity
  • Availability
  • Add to Cart