MAN372905389 Manchester Organics

2-Amino-4,6-dimethoxybenzoic acid

  • MDL number MFCD09031989
  • Cas Number : 21577-57-1
  • Molecular Weight : 197.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/R25658
Availability details for
Quantity
  • Availability
  • Add to Cart