MAN372905501 Manchester Organics

8-Bromo-2-methylquinoline

  • MDL number MFCD04966997
  • Molecular Weight : 222.081

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D23565
Availability details for
Quantity
  • Availability
  • Add to Cart