MAN372905509 Manchester Organics

Methyl 6-(trifluoromethyl)pyridine-2-carboxylate

  • MDL number MFCD02090527
  • Cas Number : 155377-05-2
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M15525
Availability details for
Quantity
  • Availability
  • Add to Cart