MAN372905520 Manchester Organics

Ethyl 2-bromo-2-cyclopropylacetate

  • MDL number MFCD18837719
  • Molecular Weight : 207.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G23568
Availability details for
Quantity
  • Availability
  • Add to Cart