MAN372905529 Manchester Organics

tert-butyl 3-Fluoro-4-hydroxypiperidine-1-carboxylate

  • MDL number MFCD11977352
  • Cas Number : 373604-28-5
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T26654
Availability details for
Quantity
  • Availability
  • Add to Cart