MAN372905573 Manchester Organics

2-Fluoro-4-iodo-6-methyl-pyridine

  • MDL number MFCD03095298
  • Cas Number : 884494-45-5
  • Molecular Weight : 237.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-4-iodo-6-methylpyridine
Inchi Code 1S/C6H5FIN/c1-4-2-5(8)3-6(7)9-4/h2-3H,1H3
Inchi Key LJTFIDJMXXZXPN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/K26670
Availability details for
Quantity
  • Availability
  • Add to Cart